missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyl orthosilicate, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC203820025

 View more versions of this product

Catalog No. AC203820025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tetramethyl orthosilicate
26°C
Glass bottle
1.03
Undesignated
2.5kg
99%
Si(OCH3)4
Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol
CO[Si](OC)(OC)OC
152.221
152.22
99%
1.0300g/mL
Authentic
1.3670 to 1.3700
121.0°C to 122.0°C
-2.0°C
681-84-5
C4H12O4Si
methyl orthosilicate
LFQCEHFDDXELDD-UHFFFAOYSA-N
tetramethyl silicate
12682
Liquid

Chemical Identifiers

681-84-5
152.221
methyl orthosilicate
tetramethyl silicate
C4H12O4Si
LFQCEHFDDXELDD-UHFFFAOYSA-N
12682
CO[Si](OC)(OC)OC
SDS Documents