Learn More
Tetramethyl orthocarbonate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6661703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetramethyl orthocarbonate | |
-5°C | |
1.017 g/mL | |
22°C (72°F) | |
1.385 | |
MFCD00008473 | |
UN3272 | |
Moisture sensitive | |
AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
tetramethoxymethane | |
74613 | |
95% |
1850-14-2 | |
Colorless | |
114°C | |
C5H12O4 | |
C(OCH3)4 | |
1 g | |
1737682 | |
tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
COC(OC)(OC)OC | |
136.147 | |
136.15 | |
Liquid |
Chemical Identifiers
1850-14-2 | |
136.147 | |
AHJWSRRHTXRLAQ-UHFFFAOYSA-N | |
74613 | |
COC(OC)(OC)OC |
C5H12O4 | |
MFCD00008473 | |
tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc | |
tetramethoxymethane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 217-438-5
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only