missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyl methylenediphosphonate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1344122

 View more versions of this product

Catalog No. AAA1344122



Description

Description

Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetramethyl methylenediphosphonate
152°C to 154°C (2.5 mmHg)
1.455
100 g
tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester
XAVFZUKFLWOSOS-UHFFFAOYSA-N
bis(dimethoxyphosphoryl)methane
519206
≥98%
16001-93-7
C5H14O6P2
MFCD00014884
1874756
Miscible with water.
COP(=O)(CP(=O)(OC)OC)OC
232.109
232.11

Chemical Identifiers

16001-93-7
232.109
XAVFZUKFLWOSOS-UHFFFAOYSA-N
519206
COP(=O)(CP(=O)(OC)OC)OC
C5H14O6P2
MFCD00014884
tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester
bis(dimethoxyphosphoryl)methane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only