Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethoxygermanium(IV)

Sol-gel intermediate

Chemical Identifiers

Chemical Name or Material Tetramethoxygermanium(IV)
CAS 992-91-6
Formula Weight 196.73g/mol
MDL Number MFCD00014881
UN Number UN2924
Synonym Germanium(IV) methoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3260806

 
alfa aesar™
32608-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Refractive Index 1.4015
Density 1.3254g/mL at 20°C
Melting Point −18°C
Boiling Point 145°C
Quantity 5g
Flash Point 40° (105°F)
Chemical Name or Material Tetramethoxygermanium(IV)
CAS 992-91-6
Physical Form Liquid
Formula Weight 196.73g/mol
MDL Number MFCD00014881
UN Number UN2924
Synonym Germanium(IV) methoxide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.