missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrakis(triphenylphosphine)platinum(0), Pt 15.2% min

Hydrosilation and oxidation

Manufacturer:  Alfa Aesar™ 1054903

 View more versions of this product

Catalog No. AA1054903



Specifications

Specifications

Tetrakis(triphenylphosphine)platinum(0)
Air sensitive
1g
C72H60P4Pt
platinum 0-tetrakis triphenylphosphine, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum; triphenylphosphane, platinum; triphenylphosphine, pt pph3 4, tetrakis triphenylphosphine platinum, tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum 0 1g
Soluble in benzene
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt]
1244.251
1244.21
Odorless
Yellow
14221-02-4
MFCD00010014
Pt 15.2% min.
SYKXNRFLNZUGAJ-UHFFFAOYSA-N
platinum;triphenylphosphane
11979705
Crystalline Powder

Chemical Identifiers

14221-02-4
1244.251
SYKXNRFLNZUGAJ-UHFFFAOYSA-N
11979705
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt]
C72H60P4Pt
MFCD00010014
platinum 0-tetrakis triphenylphosphine, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum; triphenylphosphane, platinum; triphenylphosphine, pt pph3 4, tetrakis triphenylphosphine platinum, tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum 0 1g
platinum;triphenylphosphane
Safety and Handling

Safety and Handling

EINECSNumber : 238-087-4

TSCA : Yes

storageNote1 : 2°C to 8°C

Recommended Storage : Keep cold; Store under Argon

Documents