missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(triphenylphosphine)platinum(0), 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223960050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tetrakis(triphenylphosphine)platinum(0) | |
97.5% min. | |
98% | |
C72H60P4Pt | |
MFCD00010014 | |
15,300; 16,98; 04,33; 05,34; 09,45; 10,33; 14,36; 15,30; 16,29; 17,32; 06,570; 07,357; 09,450; 12,467; 13,294; 16,317 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt] | |
1244.27 | |
1244.27 |
14221-02-4 | |
Authentic | |
Glass bottle | |
[(C6H5)3P]4Pt | |
5g | |
tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g | |
SYKXNRFLNZUGAJ-UHFFFAOYSA-N | |
platinum;triphenylphosphane | |
11979705 | |
98% |
Chemical Identifiers
14221-02-4 | |
1244.27 | |
SYKXNRFLNZUGAJ-UHFFFAOYSA-N | |
11979705 | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pt] |
C72H60P4Pt | |
MFCD00010014 | |
tetrakis triphenylphosphine platinum 0, tetrakis triphenylphosphine platinum, platinum; triphenylphosphane, pt pph3 4, platinum tetratriphenylphosphine, platinum, tetrakis triphenylphosphine-, t-4, platinum 0-tetrakis triphenylphosphine, platinum; triphenylphosphine, tetrakis triphenylphosphine platinum 0 1g | |
platinum;triphenylphosphane |
Safety and Handling
EINECSNumber : 238-087-4
RUO â Research Use Only