missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202380250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrakis(triphenylphosphine)palladium(0) | |
103.0°C to 107.0°C | |
98.5% min. | |
99% | |
C72H60P4Pd | |
MFCD00010012 | |
06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327 | |
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
palladium;triphenylphosphane | |
11979704 | |
99% |
14221-01-3 | |
Khaki to Yellow | |
Authentic | |
Glass bottle | |
[(C6H5)3P]4Pd | |
25 g | |
15,9367 | |
Solubility in water: insoluble. | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
1155.59 | |
1155.58 | |
Fine Powder or Platelets |
Chemical Identifiers
14221-01-3 | |
1155.59 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
11979704 | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C72H60P4Pd | |
MFCD00010012 | |
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 | |
palladium;triphenylphosphane |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 238-086-9
RUO – Research Use Only