missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(triphenylphosphine)palladium(0), 99,8% (metals basis), Pd 9% min, Alfa Aesar™

Hydrosilation, isomerization, carbonylation

Manufacturer:  Alfa Aesar 1054809

 View more versions of this product

Catalog No. AA1054809



Specifications

Specifications

Tetrakis(triphenylphosphine)palladium
10g
99.8% (metals basis), Pd 9% min
MFCD00010012
NFHFRUOZVGFOOS-UHFFFAOYSA-N
palladium;triphenylphosphane
11979704
Crystalline
100°C to 105°C
14221-01-3
C72H60P4Pd
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
1155.587
1155.57

Chemical Identifiers

14221-01-3
1155.587
NFHFRUOZVGFOOS-UHFFFAOYSA-N
11979704
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
C72H60P4Pd
MFCD00010012
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
palladium;triphenylphosphane
Safety and Handling

Safety and Handling

storageNote1 : Air Sensitive

Documents