Learn More
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals
Hydrosilation, isomerization, carbonylation
Supplier: Thermo Scientific Chemicals 01054804
Description
Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrakis(triphenylphosphine)palladium(0) | |
100°C to 105°C | |
C72H60P4Pd | |
2 g | |
Air sensitive | |
Insoluble in water. Soluble in benzene,ethanol and chloroform | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
1155.59 | |
1155.57 |
14221-01-3 | |
99.8% (metals basis), Pd 9% min. | |
MFCD00010012 | |
6704828 | |
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
palladium;triphenylphosphane | |
11979704 | |
Crystalline |
Chemical Identifiers
14221-01-3 | |
1155.59 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C72H60P4Pd | |
MFCD00010012 | |
11979704 |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302-H500
EINECSNumber : 238-086-9
TSCA : Yes
storageNote1 : Air Sensitive
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only