Please login to your online account to display your discounted pricing

Tetrakis(triphenylphosphine)nickel(0), 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  223980010

 View more versions of this product

Catalog No. AC223980010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Fine Powder
Formula Weight 1107.89
Quantity 1g
Color Rust-brown to brown
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C72H60NiP4
Linear Formula {(C6H5)3P}4Ni
MDL Number MFCD00010011
CAS 15133-82-1

Specifications

Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
Name Note 95%
CAS 15133-82-1
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C72H60NiP4
Linear Formula {(C6H5)3P}4Ni
Formula Weight 1107.89
MDL Number MFCD00010011
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.

SDS & Safety Documents