Please login to your online account to display your discounted pricing

Tetrakis(triphenylphosphine)nickel(0), 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
Name Note 95%
CAS 15133-82-1
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C72H60NiP4
Linear Formula {(C6H5)3P}4Ni
Formula Weight 1107.89
MDL Number MFCD00010011
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC223980010

  SDS
fisher scientific acros
223980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC223980050

  SDS
thermo fisher scientific
AC223980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 1107.89
Physical Form Fine Powder
Quantity 1g
Color Rust-brown to brown
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 95%
Name Note 95%
Linear Formula {(C6H5)3P}4Ni
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
Molecular Formula C72H60NiP4
MDL Number MFCD00010011
CAS 15133-82-1
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Suspected of causing cancer.
Flammable solid.