Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrakis(triphenylphosphine)nickel(0)

Chemical Identifiers

Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
CAS 15133-82-1
Linear Formula Ni[P(C6H5)3]4
Formula Weight 1107.83
MDL Number MFCD00010011
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3210103

  SDS
alfa aesar™
3210103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA32101MD

  SDS
alfa aesar™
32101MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1107.83
Health Hazard 3 GHS P Statement
P210-P201-P309-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)
Quantity 1g
UN Number UN1325
Linear Formula Ni[P(C6H5)3]4
MDL Number MFCD00010011
Melting Point 123° to 128°C
Sensitivity Air & Moisture Sensitive
Physical Form Crystalline
CAS 15133-82-1
Health Hazard 2 GHS H Statement
H350-H372-H228-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Flammable solid.
May cause an allergic skin reaction.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Color Brown
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P201-P309-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H350-H372-H228-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Flammable solid.
May cause an allergic skin reaction.