Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrakis(trimethylsiloxy)silane, 98+%

$21.09 - $54.85

Chemical Identifiers

Chemical Name or Material Tetrakis(trimethylsiloxy)silane
CAS 3555-47-3
Molecular Formula C12H36O4Si5
Formula Weight 384.85
MDL Number MFCD00051587
Beilstein 1793898
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0608303

  SDS
alfa aesar™
L0608303
1g Each for $21.09

AAL0608306

  SDS
alfa aesar™
L0608306
5g Each for $54.85
Description & Specifications

Specifications

Formula Weight 384.85
Assay ≥98%
Molecular Formula C12H36O4Si5
Flash Point 80°C (176°F)
CAS 3555-47-3
Chemical Name or Material Tetrakis(trimethylsiloxy)silane
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
MDL Number MFCD00051587
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 1793898
Quantity 1g
TSCA TSCA
Boiling Point 103° to 106°C (2mmHg)
Melting Point −60°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.