Please login to your online account to display your discounted pricing

Tetrakis(dimethylamino)zirconium, ACROS Organics™


Quantity 5g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC380410050

 View more versions of this product

Catalog No. AC380410050

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Pale yellow-greenish
Physical Form Crystalline Solid
Formula Weight 267.53
Quantity 5g
Packaging Glass Bottle in Tin Can
NMR Structure confirmed
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 19756-04-8
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C8H24N4Zr
Health Hazard 2 GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Chemical Name or Material Tetrakis(dimethylamino)zirconium
MDL Number MFCD00239502

Specifications

Chemical Name or Material Tetrakis(dimethylamino)zirconium
CAS 19756-04-8
Molecular Formula C8H24N4Zr
Formula Weight 267.53
MDL Number MFCD00239502
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.

SDS & Safety Documents