Please login to your online account to display your discounted pricing

Tetrakis(dimethylamino)zirconium, ACROS Organics™

$192.64 - $192.64

Chemical Identifiers

Chemical Name or Material Tetrakis(dimethylamino)zirconium
CAS 19756-04-8
Molecular Formula C8H24N4Zr
Formula Weight 267.53
MDL Number MFCD00239502
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380410010

  SDS
thermo fisher scientific
AC380410010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380410050

  SDS
thermo fisher scientific
AC380410050
Glass Bottle in Tin Can 5g Each for $192.64
Description & Specifications

Specifications

Physical Form Crystalline Solid
Formula Weight 267.53
Color Pale yellow-greenish
Quantity 1g
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Tetrakis(dimethylamino)zirconium
NMR Structure confirmed
Packaging Glass bottle
Molecular Formula C8H24N4Zr
MDL Number MFCD00239502
CAS 19756-04-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.