missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(dimethylamino)titanium(IV), TCI America™

Manufacturer:  TCI America T22305G

 View more versions of this product

Catalog No. T22305G



Specifications

Specifications

Tetrakis(dimethylamino)titanium(IV)
5g
C8H24N4Ti
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Ti+4]
224.175
224.18
≥97.0% (T)
Undesignated
3275-24-9
3399
MNWRORMXBIWXCI-UHFFFAOYSA-N
dimethylazanide;titanium(4+)
123185
Clear Liquid at 20°C

Chemical Identifiers

3275-24-9
224.175
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
dimethylazanide;titanium(4+)
C8H24N4Ti
MNWRORMXBIWXCI-UHFFFAOYSA-N
123185
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Ti+4]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents