missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(dimethylamino)titanium(IV) 97.0+%, TCI America™
Supplier: TCI America T22301G
Specifications
Tetrakis(dimethylamino)titanium(IV) | |
C8H24N4Ti | |
1 g | |
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva | |
[Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C | |
224.18 | |
224.18 | |
Liquid |
3275-24-9 | |
MFCD00014861 | |
3399 | |
MNWRORMXBIWXCI-UHFFFAOYSA-N | |
titanium(4+) tetrakis(dimethylazanide) | |
123185 | |
≥97.0% (T) |
Chemical Identifiers
3275-24-9 | |
224.18 | |
MNWRORMXBIWXCI-UHFFFAOYSA-N | |
123185 | |
[Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C |
C8H24N4Ti | |
MFCD00014861 | |
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva | |
titanium(4+) tetrakis(dimethylazanide) |
Safety and Handling
TSCA : Yes