Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
CAS 3275-24-9
Molecular Formula C8H24N4Ti
Linear Formula Ti[N(CH3)2]4
Formula Weight 224.19
MDL Number MFCD00014861
UN Number UN3129
Synonym Titanium dimethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4293206

  SDS
alfa aesar™
4293206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.19
CAS 3275-24-9
UN Number UN3129
Molecular Formula C8H24N4Ti
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Assay 99.9%
Flash Point −30°C (−22°F)
TSCA TSCA
Boiling Point 50°C (0.5mmHg)
Assay Percent Notes (metals basis)
Physical Form Liquid
Synonym Titanium dimethylamide
Health Hazard 1 Danger
Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
Linear Formula Ti[N(CH3)2]4
Health Hazard 2 GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Sensitivity Moisture Sensitive
Color Yellow-brown
Density 0.96g/mL
MDL Number MFCD00014861
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.