Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrakis(diethylamino)titanium(IV), 99.99% (metals basis)

Nitride precursor - MOCVD

Chemical Identifiers

Chemical Name or Material Tetrakis(diethylamino)titanium(IV)
CAS 4419-47-0
Linear Formula Ti[N(C2H5)2]4
Formula Weight 336.4
MDL Number MFCD00015648
UN Number UN3129
Synonym Titanium diethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4046904

  SDS
alfa aesar™
4046904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4046909

  SDS
alfa aesar™
4046909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4046918

  SDS
alfa aesar™
4046918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 336.4
Synonym Titanium diethylamide
Physical Form Liquid
TSCA TSCA
MDL Number MFCD00015648
Health Hazard 2 GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Linear Formula Ti[N(C2H5)2]4
Sensitivity Air & Moisture Sensitive
CAS 4419-47-0
Health Hazard 3 GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2g
Chemical Name or Material Tetrakis(diethylamino)titanium(IV)
Color Orange to red
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
Assay Percent Notes (metals basis)
Refractive Index 1.5370
Boiling Point 133°C (1.2mmHg)
UN Number UN3129
Density 0.95g/mL
Assay 99.99%
Safety and Handling

Danger

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.