missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetate, TCI America™

Manufacturer:  TCI America T284520MG

 View more versions of this product

Catalog No. T284520MG



Specifications

Specifications

Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetate
98°C
126150-97-8
MFCD00036696
YJIYWYAMZFVECX-UHFFFAOYSA-N
acetyloxymethyl 2-[N-[2-(acetyloxymethoxy)-2-oxoethyl]-2-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]anilino]acetate
2293
Crystal-Powder at 20°C
White
20mg
C34H40N2O18
bapta-am, bapta/am, bapta am, tetrakis acetoxymethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrakis acetoxymethyl ester, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 2-2-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy phenyl amino acetate, 1,2-bis o-aminophenoxy ethane-n,n,n′,n′-tetraacetic acid tetra acetoxymethyl ester, acmc-20d2ub, bapta, am, bapta-am hplc
CC(=O)OCOC(=O)CN(CC(=O)OCOC(=O)C)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C
764.69
764.69
≥95.0% (HPLC)

Chemical Identifiers

126150-97-8
764.69
YJIYWYAMZFVECX-UHFFFAOYSA-N
2293
CC(=O)OCOC(=O)CN(CC(=O)OCOC(=O)C)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C
C34H40N2O18
MFCD00036696
bapta-am, bapta/am, bapta am, tetrakis acetoxymethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrakis acetoxymethyl ester, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 2-2-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy phenyl amino acetate, 1,2-bis o-aminophenoxy ethane-n,n,n′,n′-tetraacetic acid tetra acetoxymethyl ester, acmc-20d2ub, bapta, am, bapta-am hplc
acetyloxymethyl 2-[N-[2-(acetyloxymethoxy)-2-oxoethyl]-2-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]anilino]acetate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents