missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(acetonitrile)copper(I) Tetrafluoroborate 98.0+%, TCI America™

Supplier:  TCI America T26665G

 View more versions of this product

Catalog No. T26665G


Description

Description

Specifications

Specifications

Tetrakis(acetonitrile)copper(I) Tetrafluoroborate
164°C
C8H12BCuF4N4
5 g
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
314.56
314.56
Crystalline Powder
15418-29-8
White
MFCD09265110
1759
YZGSKMIIVMCEFE-UHFFFAOYSA-N
λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroboranuide
10990673
≥98.0% (T)

Chemical Identifiers

15418-29-8
314.56
YZGSKMIIVMCEFE-UHFFFAOYSA-N
10990673
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
C8H12BCuF4N4
MFCD09265110
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate
λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroboranuide
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions