missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(acetonitrile)copper(I) Tetrafluoroborate, TCI America™

Manufacturer:  TCI America T266625G

 View more versions of this product

Catalog No. T266625G



Specifications

Specifications

Tetrakis(acetonitrile)copper(I) Tetrafluoroborate
164°C
15418-29-8
MFCD09265110
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate, tetrakis acetonitrile copper i tetrafluoroborate
[B-](F)(F)(F)F.CC#N.CC#N.CC#N.CC#N.[Cu+]
314.562
314.56
≥98.0% (T)
White
25g
C8H12BCuF4N4
1759
YZGSKMIIVMCEFE-UHFFFAOYSA-N
acetonitrile;copper(1+);tetrafluoroborate
10990673
Crystal-Powder at 20°C

Chemical Identifiers

15418-29-8
314.562
YZGSKMIIVMCEFE-UHFFFAOYSA-N
10990673
[B-](F)(F)(F)F.CC#N.CC#N.CC#N.CC#N.[Cu+]
C8H12BCuF4N4
MFCD09265110
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate, tetrakis acetonitrile copper i tetrafluoroborate
acetonitrile;copper(1+);tetrafluoroborate
Safety and Handling

Safety and Handling

TSCA : No

Documents