missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(4-bromophenyl)methane, TCI America™

Manufacturer:  TCI America T29601G

 View more versions of this product

Catalog No. T29601G



Specifications

Specifications

Tetrakis(4-bromophenyl)methane
300°C
105309-59-9
MFCD19688472
C1=CC(=CC=C1C(C2=CC=C(C=C2)Br)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br
636.019
636.02
≥95.0% (GC)
Yellow
1g
C25H16Br4
YBGIIZGNEOJSRF-UHFFFAOYSA-N
1-bromo-4-[tris(4-bromophenyl)methyl]benzene
11250692
Crystal-Powder at 20°C

Chemical Identifiers

105309-59-9
636.019
YBGIIZGNEOJSRF-UHFFFAOYSA-N
1-bromo-4-[tris(4-bromophenyl)methyl]benzene
C25H16Br4
MFCD19688472
11250692
C1=CC(=CC=C1C(C2=CC=C(C=C2)Br)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br
Safety and Handling

Safety and Handling

TSCA : No

Documents