missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol), TCI America™

Manufacturer:  TCI America T261025G

 View more versions of this product

Catalog No. T261025G



Specifications

Specifications

Tetrakis(2,4-pentanedionato)titanium(IV) (ca. 63% in Isopropyl Alcohol)
25g
C20H32O8Ti
1219
RYSXWUYLAWPLES-MTOQALJVSA-N
(Z)-4-hydroxypent-3-en-2-one;titanium
44629996
Clear Liquid at 20°C
Undesignated
17501-79-0
MFCD00045004
Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti]
448.335
444.30

Chemical Identifiers

17501-79-0
448.335
RYSXWUYLAWPLES-MTOQALJVSA-N
44629996
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.[Ti]
C20H32O8Ti
MFCD00045004
Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate
(Z)-4-hydroxypent-3-en-2-one;titanium
Safety and Handling

Safety and Handling

TSCA : Yes

Documents