missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 3945903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV) | |
219°C to 220°C | |
C44H80NbO8 | |
1 g | |
WYTIGJVEVROCSD-DSORRXEFSA-N | |
(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;niobium | |
72376395 | |
Powder |
41706-15-4 | |
325°C (decomposition) | |
MFCD00145379 | |
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato niobium iv, nb tmhd 4, niobium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, 4e-5-bis 3e-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy niobio oxy-2,2,6,6-tetramethylhept-4-en-3-one | |
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Nb] | |
830.022 | |
826 |
Chemical Identifiers
41706-15-4 | |
830.022 | |
WYTIGJVEVROCSD-DSORRXEFSA-N | |
72376395 | |
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Nb] |
C44H80NbO8 | |
MFCD00145379 | |
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato niobium iv, nb tmhd 4, niobium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, 4e-5-bis 3e-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy niobio oxy-2,2,6,6-tetramethylhept-4-en-3-one | |
(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;niobium |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures