Please login to your online account to display your discounted pricing

Tetrakis(1-pyrazolyl)borate, potassium salt, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrakis(1-pyrazolyl)borate, potassium salt
Name Note 95%
CAS 14782-58-2
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H12BKN8
Formula Weight 318.2
MDL Number MFCD00800557
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC300160010

  SDS
thermo fisher scientific
300160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC300160050

  SDS
thermo fisher scientific
AC300160050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Fine Crystalline Powder
Formula Weight 318.2
Quantity 1g
Color Almost white
Packaging Glass bottle
Chemical Name or Material Tetrakis(1-pyrazolyl)borate, potassium salt
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C12H12BKN8
MDL Number MFCD00800557
CAS 14782-58-2