Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraisopropyltin, 97+%

Chemical Identifiers

Chemical Name or Material Tetraisopropyltin
CAS 2949-42-0
Linear Formula Sn[(CH3)2CH]4
Formula Weight 291.05
MDL Number MFCD00026346
UN Number UN2788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3960904

  SDS
alfa aesar™
3960904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3960909

  SDS
alfa aesar™
3960909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.05
Chemical Name or Material Tetraisopropyltin
Boiling Point 112° to 114°C (18mmHg)
Physical Form Liquid
Assay ≥97%
Linear Formula Sn[(CH3)2CH]4
MDL Number MFCD00026346
CAS 2949-42-0
Quantity 2g
UN Number UN2788
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Odor Characteristic
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.124g/mL
Color Colorless
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Refractive Index 1.4851
RTECS Number WH8628000
Sensitivity Air & Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.