missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrothiopyran-4-ylmethylamine hydrochloride, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC61963CB

 View more versions of this product

Catalog No. CC61963CB

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

167.7
97%
950603-21-1
Tetrahydrothiopyran-4-ylmethylamine hydrochloride
97%
Amber Glass Bottle
250mg
100.0
MFCD09879925
C6H13NS·ClH
96.5

Chemical Identifiers

950603-21-1
100.0
C6H13NS·ClH
MFCD09879925
97%
96.5
97%
167.7
Tetrahydrothiopyran-4-ylmethylamine hydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning