Please login to your online account to display your discounted pricing

tetrahydrothiopyran-4-carbonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Tetrahydrothiopyran-4-carbonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9NS
Formula Weight 127.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC61916DA

  SDS
fisher scientific company
CC61916DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC61916DE

  SDS
fisher scientific company
CC61916DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.21
Quantity 1g
Assay 97%
Chemical Name or Material Tetrahydrothiopyran-4-carbonitrile
Packaging Amber Glass Bottle
Molecular Formula C6H9NS
CAS 195503-40-3
Assay Percent Range 97%