Please login to your online account to display your discounted pricing

tetrahydrothiopyran-4-carbonitrile, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC61916DA

 View more versions of this product

Catalog No. CC61916DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 127.21
Quantity 1g
Assay 97%
Chemical Name or Material Tetrahydrothiopyran-4-carbonitrile
Packaging Amber Glass Bottle
Molecular Formula C6H9NS
Assay Percent Range 97%
CAS 195503-40-3

Specifications

Chemical Name or Material Tetrahydrothiopyran-4-carbonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9NS
Formula Weight 127.21