Learn More
Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1390822
Description
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.
Solubility
Immiscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Tetrahydrothiophene | |
-96°C | |
120°C to 121°C | |
C4H8S | |
MFCD00005476 | |
UN2412 | |
14,9218 | |
Immiscible in water. | |
C1CCSC1 | |
88.17 | |
CHEBI:48458 | |
98% |
110-01-0 | |
0.999 | |
19°C (66°F) | |
1.504 | |
100 g | |
102392 | |
tetrahydrothiophene, thiophene, tetrahydro, thiophane, thiacyclopentane, tetramethylene sulfide, thilane, tetrahydrothiophen, pennodorant 1013, thiofan, thiolan | |
RAOIDOHSFRTOEL-UHFFFAOYSA-N | |
thiolane | |
1127 | |
88.17 |
Chemical Identifiers
110-01-0 | |
88.17 | |
RAOIDOHSFRTOEL-UHFFFAOYSA-N | |
1127 | |
thiolane |
C4H8S | |
MFCD00005476 | |
tetrahydrothiophene, thiophene, tetrahydro, thiophane, thiacyclopentane, tetramethylene sulfide, thilane, tetrahydrothiophen, pennodorant 1013, thiofan, thiolan | |
CHEBI:48458 | |
C1CCSC1 |
Safety and Handling
GHS H Statement
H225-H302-H312-H332-H315-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROTHIOPHENE
EINECSNumber : 203-728-9
RTECSNumber : XN0370000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only