Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester, 97%

$109.74 - $1,034.39

Chemical Identifiers

Chemical Name or Material Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester
Molecular Formula C8H16BNO4S
Formula Weight 233.1
MDL Number MFCD12546196
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3135503

  SDS
alfa aesar™
H3135503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3135506

  SDS
alfa aesar™
H3135506
5g Each for $1,034.39

AAH31355MD

  SDS
alfa aesar™
H31355MD
250mg Each for $109.74
Description & Specifications

Specifications

Formula Weight 233.1
Chemical Name or Material Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester
Assay 97%
Molecular Formula C8H16BNO4S
MDL Number MFCD12546196
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 242° to 244°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.