missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydropyran-4-ylacetyl chloride, 95%, Maybridge

Chemical Identifiers

CAS 40500-05-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H11ClO2
Formula Weight 162.61
MDL Number MFCD08669649
Chemical Name or Material Tetrahydropyran-4-ylacetyl chloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC67402CB SDS Fisher Scientific Company
CC67402CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67402DA SDS Fisher Scientific Company
CC67402DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

40500-05-8
100.0
C7H11ClO2
MFCD08669649
95%
94.5
95%
162.61
Tetrahydropyran-4-ylacetyl chloride

Specifications

250mg
94.5
Amber Glass Bottle
C7H11ClO2
MFCD08669649
95%
40500-05-8
100.0
95%
162.61
Tetrahydropyran-4-ylacetyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger