missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydropyran-4-ylacetaldehyde, 95%, Maybridge

Chemical Identifiers

CAS 65626-23-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD03425265
Chemical Name or Material Tetrahydropyran-4-ylacetaldehyde
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC67404CB SDS Fisher Scientific Company
CC67404CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67404DA SDS Fisher Scientific Company
CC67404DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

65626-23-5
100.0
C7H12O2
MFCD03425265
95%
94.5
95%
128.17
Tetrahydropyran-4-ylacetaldehyde

Specifications

128.17
100.0
MFCD03425265
C7H12O2
65626-23-5
95%
250mg
Amber Glass Bottle
95%
94.5
Tetrahydropyran-4-ylacetaldehyde

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning