missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydropyran-4-carboxamidine hydrochloride, 97%, Maybridge

Chemical Identifiers

CAS 426828-34-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H12N2O·ClH
Formula Weight 164.63
MDL Number MFCD10697951
Chemical Name or Material Tetrahydropyran-4-carboxamidine hydrochloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC29931CB SDS Fisher Scientific Company
CC29931CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC29931DA SDS Fisher Scientific Company
CC29931DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

426828-34-4
100.0
C6H12N2O·ClH
MFCD10697951
97%
96.5
97%
164.63
Tetrahydropyran-4-carboxamidine hydrochloride

Specifications

164.63
100.0
C6H12N2O·ClH
96.5
MFCD10697951
97%
250mg
Amber Glass Bottle
97%
426828-34-4
Tetrahydropyran-4-carboxamidine hydrochloride

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning