missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydropyran-2-carbonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 40053-81-4
Molecular Formula C6H9ClO2
Molecular Weight (g/mol) 148.586
MDL Number MFCD09025837
InChI Key CDKSDHJUTXTPHO-UHFFFAOYSA-N
Synonym 2h-3,4,5,6-tetrahydropyran-2-carbonyl chloride, 2h-pyran-2-carbonyl chloride, tetrahydro-9ci, 2h-pyran-2-carbonylchloride, tetrahydro-, r-9ci, acmc-20n6vi, tetrahydro-2h-pyran-2-carbonyl chloride, tetrahydro-pyran-2-carbonyl chloride, tetrahydropyran-2-carbonyl chloride
PubChem CID 15731743
IUPAC Name oxane-2-carbonyl chloride
SMILES C1CCOC(C1)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC41202CB
SDS View Documents Promotion Details
Fisher Scientific Company
CC41202CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

40053-81-4
148.586
CDKSDHJUTXTPHO-UHFFFAOYSA-N
15731743
C1CCOC(C1)C(=O)Cl
C6H9ClO2
MFCD09025837
2h-3,4,5,6-tetrahydropyran-2-carbonyl chloride, 2h-pyran-2-carbonyl chloride, tetrahydro-9ci, 2h-pyran-2-carbonylchloride, tetrahydro-, r-9ci, acmc-20n6vi, tetrahydro-2h-pyran-2-carbonyl chloride, tetrahydro-pyran-2-carbonyl chloride, tetrahydropyran-2-carbonyl chloride
oxane-2-carbonyl chloride

Specifications

40053-81-4
100.0
C6H9ClO2
Amber Glass Bottle
250mg
C1CCOC(C1)C(=O)Cl
148.586
148.59
Tetrahydropyran-2-carbonyl chloride
94.5
95%
MFCD09025837
2h-3,4,5,6-tetrahydropyran-2-carbonyl chloride, 2h-pyran-2-carbonyl chloride, tetrahydro-9ci, 2h-pyran-2-carbonylchloride, tetrahydro-, r-9ci, acmc-20n6vi, tetrahydro-2h-pyran-2-carbonyl chloride, tetrahydro-pyran-2-carbonyl chloride, tetrahydropyran-2-carbonyl chloride
CDKSDHJUTXTPHO-UHFFFAOYSA-N
oxane-2-carbonyl chloride
15731743
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications