Learn More
(+/-)-Tetrahydrofurfurylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2306836
Description
Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.
Solubility
Miscible with water.
Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, water/ moisture. Store under dry inert gas.
Specifications
(+/-)-Tetrahydrofurfurylamine | |
0.98 | |
500 g | |
C5H11NO | |
UN2943 | |
tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine | |
YNOGYQAEJGADFJ-UHFFFAOYNA-N | |
oxolan-2-ylmethanamine | |
253298 | |
97% |
4795-29-3 | |
153°C to 154°C | |
1.455 | |
MFCD00005373 | |
102729 | |
Miscible with water. | |
NCC1CCCO1 | |
101.15 | |
101.15 |
Chemical Identifiers
4795-29-3 | |
101.15 | |
YNOGYQAEJGADFJ-UHFFFAOYNA-N | |
253298 | |
NCC1CCCO1 |
C5H11NO | |
MFCD00005373 | |
tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine | |
oxolan-2-ylmethanamine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAHYDROFURFURYLAMINE
EINECSNumber : 225-351-9
RTECSNumber : LV0175000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only