Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofurfuryl methacrylate, 97%, stab.

$26.36 - $58.15

Chemical Identifiers

Chemical Name or Material Tetrahydrofurfuryl methacrylate
CAS 2455-24-5
Name Note stabilized
Molecular Formula C9H14O3
Formula Weight 170.21
MDL Number MFCD00014515
Beilstein 117423
Synonym Methacrylic acid tetrahydrofurfuryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0936518

  SDS
alfa aesar™
L0936518
50g Each for $26.36

AAL0936530

  SDS
alfa aesar™
L0936530
250g Each for $58.15
Description & Specifications

Specifications

Formula Weight 170.21
Chemical Name or Material Tetrahydrofurfuryl methacrylate
CAS 2455-24-5
MDL Number MFCD00014515
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Beilstein 117423
Recommended Storage 2° to 8°C
Molecular Formula C9H14O3
Synonym Methacrylic acid tetrahydrofurfuryl ester
Health Hazard 3 GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Quantity 50g
TSCA TSCA
Boiling Point 178°C
Flash Point 90°C (194°F)
Assay 97%
Health Hazard 1 Warning
Name Note stabilized
Safety and Handling

Warning

GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C