Please login to your online account to display your discounted pricing

Tetrahydrofurfuryl chloride, 98+%, ACROS Organics™

$212.61 - $212.61

Chemical Identifiers

Chemical Name or Material Tetrahydrofurfuryl chloride
Name Note 98+%
CAS 3003-84-7
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C5H9ClO
Formula Weight 120.58g/mol
MDL Number MFCD00005370
Synonym 2-(Chloromethyl)tetrahydrofuran
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC279505000

  SDS
thermo fisher scientific
279505000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC279500250

  SDS
thermo fisher scientific
279500250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC279501000

  SDS
thermo fisher scientific
279501000
Glass bottle 100g Each for $212.61
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Density 1.1100g/mL
Color Clear colorless to light yellow
Quantity 500g
Chemical Name or Material Tetrahydrofurfuryl chloride
Name Note 98+%
CAS 3003-84-7
Physical Form Clear colorless to light yellow liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C5H9ClO
Formula Weight 120.58g/mol
MDL Number MFCD00005370
Beilstein 17, III, 61
Synonym 2-(Chloromethyl)tetrahydrofuran
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number LU0350000
EINECS Number 221-102-3
TSCA TSCA
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.