Please login to your online account to display your discounted pricing

Tetrahydrofurfuryl chloride, 98+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  279501000

 View more versions of this product

Catalog No. AC279501000

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Density 1.1100g/mL
Color Clear colorless to light yellow
Quantity 100g
Chemical Name or Material Tetrahydrofurfuryl chloride
Name Note 98+%
CAS 3003-84-7
Physical Form Clear colorless to light yellow liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C5H9ClO
Formula Weight 120.58g/mol
MDL Number MFCD00005370
Beilstein 17, III, 61
Synonym 2-(Chloromethyl)tetrahydrofuran
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number LU0350000
EINECS Number 221-102-3
TSCA TSCA

Specifications

Chemical Name or Material Tetrahydrofurfuryl chloride
Name Note 98+%
CAS 3003-84-7
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C5H9ClO
Formula Weight 120.58g/mol
MDL Number MFCD00005370
Synonym 2-(Chloromethyl)tetrahydrofuran
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents