Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofurfuryl chloride, 98%

Chemical Identifiers

Chemical Name or Material Tetrahydrofurfuryl chloride
CAS 3003-84-7
Molecular Formula C5H9ClO
Formula Weight 120.58
MDL Number MFCD00005370
UN Number UN1993
Beilstein 102716
Synonym 2-(Chloromethyl)tetrahydrofuran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2401514

  SDS
alfa aesar™
B2401514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2401522

  SDS
alfa aesar™
B2401522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.58
Chemical Name or Material Tetrahydrofurfuryl chloride
TSCA TSCA
Assay 98%
Molecular Formula C5H9ClO
MDL Number MFCD00005370
CAS 3003-84-7
UN Number UN1993
Beilstein 102716
Synonym 2-(Chloromethyl)tetrahydrofuran
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 150° to 151°C
Flash Point 47°C (116°F)
Quantity 25g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number LU0350000

Applications
Tetrahydrofurfuryl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and alkali metals. It is sensitive to moisture.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.