missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofurfuryl bromide, 95%

Manufacturer:  Alfa Aesar™ A1519230

 View more versions of this product

Catalog No. AAA1519230



Specifications

Specifications

Tetrahydrofurfuryl bromide
169°C to 171°C
1192-30-9
MFCD00016894
tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, 2-bromomethyl-tetrahydrofuran, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
C1CC(OC1)CBr
165.03
165.04
61°C (142°F)
250g
C5H9BrO
102717
VOHILFSOWRNVJJ-UHFFFAOYSA-N
2-(bromomethyl)oxolane
14501
95%

Chemical Identifiers

1192-30-9
165.03
VOHILFSOWRNVJJ-UHFFFAOYSA-N
14501
C1CC(OC1)CBr
C5H9BrO
MFCD00016894
tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, 2-bromomethyl-tetrahydrofuran, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
2-(bromomethyl)oxolane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LU6000000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents