Please login to your online account to display your discounted pricing

Tetrahydrofurfuryl Alcohol, +99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  420440050

 View more versions of this product

Catalog No. AC420440050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 102.13
Quantity 5g
Color Clear
TSCA TSCA
EINECS Number 202-625-6
RTECS Number LU2450000
Density 1.0543g/mL
Beilstein 17, V,3, 115
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Synonym THFA
Chemical Name or Material Tetrahydrofurfuryl alcohol
Assay Percent Range 99+%
MDL Number MFCD00005372
Health Hazard 1 HYGROSCOPIC
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99+%
Water 0.3% max.
Molecular Formula C5H10O2
CAS 97-99-4
Furfuryl Alcohol furfuryl alcohol: < 0.1%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.

Specifications

Chemical Name or Material Tetrahydrofurfuryl alcohol
Name Note 99+%
CAS 97-99-4
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H10O2
Formula Weight 102.13
MDL Number MFCD00005372
Synonym THFA
Accessories

HYGROSCOPIC

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents