Learn More
Tetrahydrofurfuryl acrylate, tech. 90%, stab., Thermo Scientific Chemicals
$63.79 - $219.96
Chemical Identifiers
CAS | 2399-48-6 |
---|---|
Molecular Formula | C8H12O3 |
Molecular Weight (g/mol) | 156.181 |
MDL Number | MFCD00014516 |
InChI Key | YNXCGLKMOXLBOD-UHFFFAOYSA-N |
Synonym | tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester |
PubChem CID | 94232 |
IUPAC Name | oxolan-2-ylmethyl prop-2-enoate |
SMILES | C=CC(=O)OCC1CCCO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1029918
|
Thermo Scientific Chemicals
L1029918 |
50 g |
N/A
|
|
|||||
AAL1029930
|
Thermo Scientific Chemicals
L1029930 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2399-48-6 | |
156.181 | |
YNXCGLKMOXLBOD-UHFFFAOYSA-N | |
94232 | |
C=CC(=O)OCC1CCCO1 |
C8H12O3 | |
MFCD00014516 | |
tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester | |
oxolan-2-ylmethyl prop-2-enoate |
Specifications
2399-48-6 | |
88°C to 89°C (9 mmHg) | |
C8H12O3 | |
MFCD00014516 | |
UN3334 | |
tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester | |
C=CC(=O)OCC1CCCO1 | |
156.181 | |
156.18 | |
Technical |
1.03 | |
93°C (199°F) | |
1.455 | |
50 g | |
113115 | |
YNXCGLKMOXLBOD-UHFFFAOYSA-N | |
oxolan-2-ylmethyl prop-2-enoate | |
94232 | |
90% | |
Tetrahydrofurfuryl acrylate, stabilized with 500ppm hydroquinone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H314-H317-H360Df
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber : 219-268-7
RTECSNumber : UD3670893
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only