Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrahydrofurfuryl acetate, 98%

$38.42 - $145.26

Chemical Identifiers

Chemical Name or Material Tetrahydrofurfuryl acetate
CAS 637-64-9
Molecular Formula C7H12O3
Formula Weight 144.17
MDL Number MFCD00022478
Beilstein 383629
Synonym Acetic acid tetrahydrofurfuryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2320518

  SDS
alfa aesar™
B2320518
50g Each for $38.42

AAB2320530

  SDS
alfa aesar™
B2320530
250g Each for $145.26
Description & Specifications

Specifications

Formula Weight 144.17
Synonym Acetic acid tetrahydrofurfuryl ester
MDL Number MFCD00022478
Flash Point 81°C (177°F)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H12O3
Quantity 50g
Beilstein 383629
TSCA TSCA
CAS 637-64-9
Health Hazard 1 Warning
Boiling Point 193° to 195°C
Assay 98%
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Chemical Name or Material Tetrahydrofurfuryl acetate
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.