missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, UV, >99.7% min, HPLC Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 22904K2

 View more versions of this product

Catalog No. AA22904K2



Specifications

Specifications

Tetrahydrofuran
109-99-9
≥99.7%
MFCD00005356
102391
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
0.889
1.4070
66°C
-108°C
unstab.
HPLC Grade
C4H8O
UN2056
14,9211
C1CCOC1
72.107
CHEBI:26911
Liquid
Ether-like
−17°C (1°F)
Colorless
1L

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
CHEBI:26911
C1CCOC1
C4H8O
MFCD00005356
8028
oxolane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H351-H319-H335;
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN

RTECSNumber : LU5950000

TSCA : TSCA

SDS Documents