Please login to your online account to display your discounted pricing

Tetrahydrofuran, UV, HPLC Grade, 99.7+%, unstab., Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Name Note unstab.
Purity Grade HPLC Grade
Molecular Formula C4H8O
Formula Weight 72.11
MDL Number MFCD00005356
UN Number UN2056
Beilstein 102391
Merck Index 14,9211
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA22904K2

 
alfa aesar
22904K2
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 72.11
Quantity 1L
Chemical Name or Material Tetrahydrofuran
TSCA TSCA
Odor Ether-like
Refractive Index 1.4070
Name Note unstab.
Purity Grade HPLC Grade
Flash Point −17°C (1°F)
Boiling Point 66°C
Physical Form Liquid
Color Colorless
Melting Point −108°C
Assay ≥99.7%
Molecular Formula C4H8O
MDL Number MFCD00005356
CAS 109-99-9
UN Number UN2056
Beilstein 102391
Merck Index 14,9211
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H351-H319-H335;
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Density 0.889
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
RTECS Number LU5950000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H351-H319-H335;
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.