missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrofuran, UV, >99.7% min, HPLC Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 22904AK

 View more versions of this product

Catalog No. AA22904AK



Specifications

Specifications

Tetrahydrofuran
109-99-9
Colorless liquid
C4H8O
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
C1CCOC1
72.107
CHEBI:26911
Liquid
Ether-like
Colorless
Unstable
HPLC, UV
>99.7%
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
0.889g/cm3 at 20°C
200 hPa (150mm Hg) at 20°C
250mL

Chemical Identifiers

109-99-9
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1
Safety and Handling

Safety and Handling

H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-726-8

SDS Documents