Please login to your online account to display your discounted pricing

Tetrahydrofuran, Ultra Low water, BakerDRY™, J.T.Baker™

Specifically manufactured for use in organometallic and oligonucleotide synthesis

$299.07 - $472.44

Chemical Identifiers

Chemical Name or Material Tetrahydrofuran
CAS 109-99-9
Name Note Ultra Low water
Purity Grade BakerDRY™
Molecular Formula C4H8O
Formula Weight 72.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004430

 
avantor performance
944710
Glass Bottle 100mL Each for $299.07

02004431

 
avantor performance
944712
Narrow Mouth Amber Glass Bottle, w/ Septum 1L Each for $472.44
Description & Specifications

Specifications

Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Packaging Glass Bottle
Specific Gravity 1L = 0.88kg
Name Note Ultra Low water
Purity Grade BakerDRY™
Flash Point −14.00°C
Quantity 100mL
Molecular Formula C4H8O
CAS 109-99-9

    For Use in Organic Synthesis, including Organometallic Synthesis