Learn More
Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 41819AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrahydrofuran | |
Unstable | |
Colorless | |
0.889 g/cm3 at 20°C | |
250 mL | |
Ether-like | |
1.407 | |
C4H8O | |
200 hPa (150mm Hg) at 20°C | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 |
°C to 108°C | |
109-99-9 | |
Colorless liquid | |
66°C | |
-17°C (1°F) | |
Liquid | |
>99.7% | |
MFCD00005356 | |
102391 | |
Miscible with water. | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
Spectrophotometric |
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302-H319-H335-H336-H351
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-726-8
RTECSNumber : LU5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only